5-Chloro-2-fluoro-3-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

56408-500MG 56408-1G 56408-5G 56408-25G
77329-678EA 168.92 USD
77329-678 77346-278 77365-958 77387-808
5-Chloro-2-fluoro-3-iodobenzaldehyde ≥97%
5-Chloro-2-fluoro-3-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃ClFIO
Storage Temperature: Ambient
MDL Number: MFCD34564362
CAS Number: 2383822-81-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR