1-(5-Chloro-2-fluoro-3-iodophenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

56405-500MG 56405-1G 56405-5G 56405-25G
77329-676EA 126.69 USD
77329-676 77344-140 77365-956 77377-850
1-(5-Chloro-2-fluoro-3-iodophenyl)ethanone ≥97%
1-(5-Chloro-2-fluoro-3-iodophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₅ClFIO
MW: 298.481 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564361
CAS Number: 2386361-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR