(5-Chloro-2-fluoro-3-methylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

56398-500MG 56398-1G 56398-5G
77335-688EA 363.59 USD
77335-688 77353-626 77377-848
(5-Chloro-2-fluoro-3-methylphenyl)methanol ≥97%
(5-Chloro-2-fluoro-3-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₈ClFO
Storage Temperature: Ambient
MDL Number: MFCD28751973
CAS Number: 1532626-99-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR