Methyl 5-chloro-2-fluoro-3-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

56397-500MG 56397-1G 56397-5G 56397-25G
77329-672EA 206 USD
77329-672 77353-624 77365-952 77387-804
Methyl 5-chloro-2-fluoro-3-methylbenzoate ≥97%
Methyl 5-chloro-2-fluoro-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₈ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD24115506
CAS Number: 1526817-07-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR