5-Chloro-2-fluoro-1-iodo-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

56395-1G 56395-5G 56395-25G
77340-742EA 60.77 USD
77340-742 77365-950 77377-846
5-Chloro-2-fluoro-1-iodo-3-methylbenzene ≥97%
5-Chloro-2-fluoro-1-iodo-3-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅ClFI
MW: 270.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28751673
CAS Number: 1805523-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR