(5-Chloro-2-fluoro-3-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

56394-1G 56394-5G 56394-25G
77341-682EA 99.91 USD
77341-682 77365-948 77377-844
(5-Chloro-2-fluoro-3-methylphenyl)(methyl)sulfane ≥97%
(5-Chloro-2-fluoro-3-methylphenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₈H₈ClFS
MW: 190.666 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28752243
CAS Number: 1805115-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR