2-(5-Chloro-2-fluoro-3-methylphenyl)propan-2-ol ≥97%

Supplier: AOBChem USA

Warning

56390-500MG 56390-1G 56390-5G 56390-25G
77329-666EA 211.15 USD
77329-666 77353-620 77365-944 77387-800
2-(5-Chloro-2-fluoro-3-methylphenyl)propan-2-ol ≥97%
2-(5-Chloro-2-fluoro-3-methylphenyl)propan-2-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₂ClFO
MW: 202.653 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28526651
CAS Number: 1780314-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR