Ethyl 5-chloro-2-fluoro-3-(methylthio)benzoate ≥97%

Supplier: AOBChem USA

Warning

56387-500MG 56387-1G 56387-5G
77338-762EA 517.06 USD
77338-762 77353-614 77377-836
Ethyl 5-chloro-2-fluoro-3-(methylthio)benzoate ≥97%
Ethyl 5-chloro-2-fluoro-3-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀ClFO₂S
Storage Temperature: Ambient
MDL Number: MFCD34757800
CAS Number: 2755717-51-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR