(5-Chloro-2-fluoro-3-(methylthio)phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

56384-500MG 56384-1G 56384-5G
77340-334EA 908.46 USD
77340-334 77353-608 77377-830
(5-Chloro-2-fluoro-3-(methylthio)phenyl)methanol ≥97%
(5-Chloro-2-fluoro-3-(methylthio)phenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₈ClFOS
MW: 206.665 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757798
CAS Number: 2764729-54-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR