2-(4-Chloro-3,5-dimethylphenyl)propan-2-ol ≥97%

Supplier: AOBChem USA

Warning

56378-500MG 56378-1G 56378-5G 56378-25G
77329-660EA 233.81 USD
77329-660 77353-606 77365-942 77387-798
2-(4-Chloro-3,5-dimethylphenyl)propan-2-ol ≥97%
2-(4-Chloro-3,5-dimethylphenyl)propan-2-ol

Precaución: for Research Use Only

Formula: C₁₁H₁₅ClO
MW: 198.689 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20528518
CAS Number: 1378860-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR