1-Ethoxy-2-fluoro-3-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

56377-500MG 56377-1G 56377-5G
77329-658EA 307.97 USD
77329-658 77353-604 77377-828
1-Ethoxy-2-fluoro-3-iodobenzene ≥97%
1-Ethoxy-2-fluoro-3-iodobenzene

Precaución: for Research Use Only

Formula: C₈H₈FIO
MW: 266.051 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530518
CAS Number: 2734777-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR