(3-Ethoxy-2-fluorophenyl)(methyl)sulphane ≥97%

Supplier: AOBChem USA

Warning

56376-1G 56376-5G 56376-25G
77340-740EA 60.77 USD
77340-740 77365-940 77377-826
(3-Ethoxy-2-fluorophenyl)(methyl)sulphane ≥97%
(3-Ethoxy-2-fluorophenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₉H₁₁FOS
Storage Temperature: Ambient
MDL Number: MFCD34530517
CAS Number: 2967402-05-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR