1,4-Difluoro-2-iodo-5-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

56373-1G 56373-5G 56373-25G
77341-036EA 66.95 USD
77341-036 77365-938 77377-824
1,4-Difluoro-2-iodo-5-methoxybenzene ≥97%
1,4-Difluoro-2-iodo-5-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₅F₂IO
MW: 270.015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530516
CAS Number: 1314894-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR