1,4-Difluoro-2-iodo-5-(methoxymethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

56371-500MG 56371-1G 56371-5G 56371-25G
77329-654EA 147.29 USD
77329-654 77345-270 77365-934 77387-796
1,4-Difluoro-2-iodo-5-(methoxymethoxy)benzene ≥95%
1,4-Difluoro-2-iodo-5-(methoxymethoxy)benzene

Precaución: for Research Use Only

Formula: C₈H₇F₂IO₂
MW: 300.041 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564357
CAS Number: 2734775-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR