6-Chloro-2-fluoro-3-iodophenol ≥95%

Supplier: AOBChem USA

Warning

56363-500MG 56363-1G 56363-5G 56363-25G
77329-652EA 170.98 USD
77329-652 77346-376 77365-932 77387-794
6-Chloro-2-fluoro-3-iodophenol ≥95%
6-Chloro-2-fluoro-3-iodophenol

Precaución: for Research Use Only

Formula: C₆H₃ClFIO
MW: 272.443 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757794
CAS Number: 2384343-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR