1-Bromo-5-chloro-4-ethoxy-2-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

56359-500MG 56359-1G 56359-5G 56359-25G
77329-650EA 260.59 USD
77329-650 77353-600 77365-930 77387-792
1-Bromo-5-chloro-4-ethoxy-2-methoxybenzene ≥97%
1-Bromo-5-chloro-4-ethoxy-2-methoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrClO₂
MW: 265.531 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757792
CAS Number: 2386990-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR