1-(Benzyloxy)-4-bromo-2-chloro-5-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

56358-500MG 56358-1G 56358-5G 56358-25G
77329-648EA 150.38 USD
77329-648 77345-356 77365-928 77387-790
1-(Benzyloxy)-4-bromo-2-chloro-5-methoxybenzene ≥97%
1-(Benzyloxy)-4-bromo-2-chloro-5-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrClO₂
MW: 327.601 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757791
CAS Number: 2901082-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR