(2-Chloro-6-(trifluoromethyl)pyridin-3-yl)(thiomorpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

56343-500MG 56343-1G 56343-5G 56343-25G
77329-646EA 139.05 USD
77329-646 77344-670 77365-926 77387-788
(2-Chloro-6-(trifluoromethyl)pyridin-3-yl)(thiomorpholino)methanone ≥97%
(2-Chloro-6-(trifluoromethyl)pyridin-3-yl)(thiomorpholino)methanone

Precaución: for Research Use Only

Formula: C₁₁H₁₀ClF₃N₂OS
Storage Temperature: Ambient
MDL Number: MFCD34757788
CAS Number: 2901066-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR