(2-Chloro-6-(trifluoromethyl)pyridin-3-yl)(morpholino)methanone ≥95%

Supplier: AOBChem USA

Warning

56342-500MG 56342-1G 56342-5G 56342-25G
77329-644EA 157.59 USD
77329-644 77345-648 77365-924 77387-786
(2-Chloro-6-(trifluoromethyl)pyridin-3-yl)(morpholino)methanone ≥95%
(2-Chloro-6-(trifluoromethyl)pyridin-3-yl)(morpholino)methanone

Precaución: for Research Use Only

Formula: C₁₁H₁₀ClF₃N₂O₂
MW: 294.658 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757787
CAS Number: 2807440-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR