(2-Chloro-6-(trifluoromethyl)pyridin-3-yl)(piperidin-1-yl)methanone ≥95%

Supplier: AOBChem USA

Warning

56341-500MG 56341-1G 56341-5G 56341-25G
77329-642EA 157.59 USD
77329-642 77345-646 77365-922 77387-784
(2-Chloro-6-(trifluoromethyl)pyridin-3-yl)(piperidin-1-yl)methanone ≥95%
(2-Chloro-6-(trifluoromethyl)pyridin-3-yl)(piperidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₂ClF₃N₂O
Storage Temperature: Ambient
MDL Number: MFCD34757786
CAS Number: 2807473-95-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR