2-Chloro-N,N-diethyl-6-(trifluoromethyl)nicotinamide ≥97%

Supplier: AOBChem USA

Warning

56340-500MG 56340-1G 56340-5G 56340-25G
77329-640EA 162.74 USD
77329-640 77346-090 77365-920 77387-782
2-Chloro-N,N-diethyl-6-(trifluoromethyl)nicotinamide ≥97%
2-Chloro-N,N-diethyl-6-(trifluoromethyl)nicotinamide

Precaución: for Research Use Only

Formula: C₁₁H₁₂ClF₃N₂O
Storage Temperature: Ambient
MDL Number: MFCD34757785
CAS Number: 2901082-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR