2-Chloro-N-methyl-6-(trifluoromethyl)nicotinamide ≥95%

Supplier: AOBChem USA

Warning

56338-500MG 56338-1G 56338-5G 56338-25G
77329-636EA 281.19 USD
77329-636 77353-594 77365-918 77387-780
2-Chloro-N-methyl-6-(trifluoromethyl)nicotinamide ≥95%
2-Chloro-N-methyl-6-(trifluoromethyl)nicotinamide

Precaución: for Research Use Only

Formula: C₈H₆ClF₃N₂O
MW: 238.594 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32877286
CAS Number: 2806436-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR