2-Chloro-N-cyclohexyl-6-(trifluoromethyl)nicotinamide ≥97%

Supplier: AOBChem USA

Warning

56336-500MG 56336-1G 56336-5G 56336-25G
77329-634EA 118.45 USD
77329-634 77343-558 77365-916 77377-814
2-Chloro-N-cyclohexyl-6-(trifluoromethyl)nicotinamide ≥97%
2-Chloro-N-cyclohexyl-6-(trifluoromethyl)nicotinamide

Precaución: for Research Use Only

Formula: C₁₃H₁₄ClF₃N₂O
MW: 306.711 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757783
CAS Number: 2901066-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR