5-Chloro-3-fluoro-2-methoxy-4'-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

56335-500MG 56335-1G 56335-5G
77336-242EA 384.19 USD
77336-242 77353-592 77377-812
5-Chloro-3-fluoro-2-methoxy-4'-methyl-1,1'-biphenyl ≥97%
5-Chloro-3-fluoro-2-methoxy-4'-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₂ClFO
MW: 250.696 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530513
CAS Number: 2734773-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR