2-Fluoro-5-(methylthio)-4-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

56329-500MG 56329-1G 56329-5G
77340-098EA 754.99 USD
77340-098 77353-590 77377-810
2-Fluoro-5-(methylthio)-4-(trifluoromethyl)benzaldehyde ≥97%
2-Fluoro-5-(methylthio)-4-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₆F₄OS
MW: 238.202 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757780
CAS Number: 2764730-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR