3-Bromo-2-chloro-5-isopropylphenol ≥97%

Supplier: AOBChem USA

Warning

56325-500MG 56325-1G 56325-5G
77338-098EA 464.53 USD
77338-098 77353-588 77377-808
3-Bromo-2-chloro-5-isopropylphenol ≥97%
3-Bromo-2-chloro-5-isopropylphenol

Precaución: for Research Use Only

Formula: C₉H₁₀BrClO
Storage Temperature: Ambient
MDL Number: MFCD34564353
CAS Number: 2734777-96-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR