3-Bromo-2,5-dimethylthiophene ≥97%

Supplier: AOBChem USA

Warning

56287-500MG 56287-1G 56287-5G 56287-25G
77329-630EA 122.57 USD
77329-630 77344-000 77365-910 77377-804
3-Bromo-2,5-dimethylthiophene ≥97%
3-Bromo-2,5-dimethylthiophene

Precaución: for Research Use Only

Formula: C₆H₇BrS
MW: 191.088 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18451735
CAS Number: 31819-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR