Methyl 4-chloro-2-methoxy-3-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

56278-500MG 56278-1G 56278-5G
77339-998EA 708.64 USD
77339-998 77353-582 77377-800
Methyl 4-chloro-2-methoxy-3-methylbenzoate ≥97%
Methyl 4-chloro-2-methoxy-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₃
MW: 214.646 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757753
CAS Number: 198344-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR