1-(4-Fluoro-3-iodophenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

56271-500MG 56271-1G 56271-5G
77336-492EA 388.31 USD
77336-492 77353-576 77377-794
1-(4-Fluoro-3-iodophenyl)ethanone ≥97%
1-(4-Fluoro-3-iodophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₆FIO
Storage Temperature: Ambient
MDL Number: MFCD21340327
CAS Number: 1824096-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR