(2-Bromo-5-iodophenyl)(4-methylpiperazin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

56239-500MG 56239-1G 56239-5G 56239-25G
77329-624EA 257.5 USD
77329-624 77353-570 77365-906 77387-774
(2-Bromo-5-iodophenyl)(4-methylpiperazin-1-yl)methanone ≥97%
(2-Bromo-5-iodophenyl)(4-methylpiperazin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₄BrIN₂O
MW: 409.061 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757750
CAS Number: 1989458-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR