(2-Bromo-5-iodophenyl)(piperidin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

56236-500MG 56236-1G 56236-25G 56236-5G
77329-620EA 300.76 USD
77329-620 77353-566 77387-770 77377-790
(2-Bromo-5-iodophenyl)(piperidin-1-yl)methanone ≥97%
(2-Bromo-5-iodophenyl)(piperidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₃BrINO
MW: 394.046 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32690494
CAS Number: 1852671-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR