(2-Bromo-5-iodophenyl)(pyrrolidin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

56234-500MG 56234-1G 56234-5G 56234-25G
77329-616EA 257.5 USD
77329-616 77353-562 77365-900 77387-766
(2-Bromo-5-iodophenyl)(pyrrolidin-1-yl)methanone ≥97%
(2-Bromo-5-iodophenyl)(pyrrolidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrINO
MW: 380.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32690493
CAS Number: 1871002-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR