2-Bromo-N-cyclohexyl-5-iodobenzamide ≥97%

Supplier: AOBChem USA

Warning

56233-500MG 56233-1G 56233-5G 56233-25G
77329-614EA 257.5 USD
77329-614 77353-560 77365-898 77387-764
2-Bromo-N-cyclohexyl-5-iodobenzamide ≥97%
2-Bromo-N-cyclohexyl-5-iodobenzamide

Precaución: for Research Use Only

Formula: C₁₃H₁₅BrINO
MW: 408.073 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757747
CAS Number: 1912043-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR