2-Bromo-N-cyclopropyl-5-iodobenzamide ≥97%

Supplier: AOBChem USA

Warning

56232-500MG 56232-1G 56232-5G 56232-25G
77329-612EA 257.5 USD
77329-612 77353-558 77365-896 77387-762
2-Bromo-N-cyclopropyl-5-iodobenzamide ≥97%
2-Bromo-N-cyclopropyl-5-iodobenzamide

Precaución: for Research Use Only

Formula: C₁₀H₉BrINO
Storage Temperature: Ambient
MDL Number: MFCD32690492
CAS Number: 1696557-00-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR