2-Bromo-5-iodo-N,N-dimethylbenzamide ≥97%
Supplier: AOBChem USA
|
Warning
|
56231-500MG
56231-1G
56231-5G
56231-25G
77329-610EA
258.53
USD
77329-610
77353-556
77365-894
77387-760
2-Bromo-5-iodo-N,N-dimethylbenzamide ≥97%
2-Bromo-5-iodo-N,N-dimethylbenzamide
Precaución: for Research Use Only
Formula:
C₉H₉BrINO MW: 353.982 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29094015 CAS Number: 1369895-41-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...