2-Bromo-5-iodo-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

56230-500MG 56230-1G 56230-25G 56230-5G
77329-608EA 300.76 USD
77329-608 77353-554 77387-758 77377-788
2-Bromo-5-iodo-N-methylbenzamide ≥97%
2-Bromo-5-iodo-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₇BrINO
MW: 339.956 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757746
CAS Number: 1694295-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR