2,2-Dichloro-1-(2-fluoro-3-methoxyphenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

56216-500MG 56216-1G 56216-5G 56216-25G
77329-606EA 189.52 USD
77329-606 77353-552 77365-892 77387-756
2,2-Dichloro-1-(2-fluoro-3-methoxyphenyl)ethanol ≥95%
2,2-Dichloro-1-(2-fluoro-3-methoxyphenyl)ethanol

Precaución: for Research Use Only

Formula: C₉H₉Cl₂FO₂
MW: 239.071 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530506
CAS Number: 2734779-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR