2,2-Dichloro-1-(3-fluoro-5-(trifluoromethyl)phenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

56215-500MG 56215-1G 56215-5G 56215-25G
77329-604EA 189.52 USD
77329-604 77353-550 77365-890 77387-754
2,2-Dichloro-1-(3-fluoro-5-(trifluoromethyl)phenyl)ethanol ≥97%
2,2-Dichloro-1-(3-fluoro-5-(trifluoromethyl)phenyl)ethanol

Precaución: for Research Use Only

Formula: C₉H₆Cl₂F₄O
MW: 277.043 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530505
CAS Number: 2734778-94-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR