1-(3-Bromo-5-methoxyphenyl)-2,2-dichloroethanol ≥95%

Supplier: AOBChem USA

Warning

56214-500MG 56214-1G 56214-5G 56214-25G
77329-602EA 185.4 USD
77329-602 77347-362 77365-888 77387-752
1-(3-Bromo-5-methoxyphenyl)-2,2-dichloroethanol ≥95%
1-(3-Bromo-5-methoxyphenyl)-2,2-dichloroethanol

Precaución: for Research Use Only

Formula: C₉H₉BrCl₂O₂
MW: 299.977 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530504
CAS Number: 2734774-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR