2,2-Dichloro-1-(3,4-dimethoxyphenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

56210-500MG 56210-1G 56210-5G 56210-25G
77329-600EA 140.08 USD
77329-600 77344-754 77365-886 77387-750
2,2-Dichloro-1-(3,4-dimethoxyphenyl)ethanol ≥95%
2,2-Dichloro-1-(3,4-dimethoxyphenyl)ethanol

Precaución: for Research Use Only

Formula: C₁₀H₁₂Cl₂O₃
MW: 251.106 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564347
CAS Number: 99094-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR