2,2-Dichloro-1-(2-chloro-3-(trifluoromethyl)phenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

56208-500MG 56208-1G 56208-5G 56208-25G
77329-598EA 189.52 USD
77329-598 77353-548 77365-884 77387-748
2,2-Dichloro-1-(2-chloro-3-(trifluoromethyl)phenyl)ethanol ≥95%
2,2-Dichloro-1-(2-chloro-3-(trifluoromethyl)phenyl)ethanol

Precaución: for Research Use Only

Formula: C₉H₆Cl₃F₃O
MW: 293.498 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530501
CAS Number: 2734773-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR