5-Chloro-2-fluoro-3-(methylthio)phenol ≥97%

Supplier: AOBChem USA

Warning

56198-500MG 56198-1G 56198-5G 56198-25G
77329-594EA 141.11 USD
77329-594 77344-838 77365-880 77387-744
5-Chloro-2-fluoro-3-(methylthio)phenol ≥97%
5-Chloro-2-fluoro-3-(methylthio)phenol

Precaución: for Research Use Only

Formula: C₇H₆ClFOS
MW: 192.638 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757743
CAS Number: 2709842-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR