5-Chloro-2-fluoro-3-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

56192-500MG 56192-1G 56192-5G
77339-368EA 582.98 USD
77339-368 77353-542 77377-786
5-Chloro-2-fluoro-3-hydroxybenzaldehyde ≥97%
5-Chloro-2-fluoro-3-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄ClFO₂
MW: 174.557 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530492
CAS Number: 2734779-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR