5-Chloro-2-fluoro-3-hydroxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

56189-500MG 56189-1G 56189-5G
77337-918EA 451.14 USD
77337-918 77353-540 77377-784
5-Chloro-2-fluoro-3-hydroxybenzoic acid ≥97%
5-Chloro-2-fluoro-3-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄ClFO₃
MW: 190.556 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31631404
CAS Number: 2386764-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR