2-Fluoro-3-formyl-6-methoxybenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
56180-500MG
56180-1G
56180-25G
56180-5G
77329-586EA
305.91
USD
77329-586
77353-534
77387-738
77377-780
2-Fluoro-3-formyl-6-methoxybenzoic acid ≥97%
2-Fluoro-3-formyl-6-methoxybenzoic acid
Precaución: for Research Use Only
Formula:
C₉H₇FO₄ MW: 198.148 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34757732 CAS Number: 2168022-69-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...