2-Chloro-4'-methyl-6-(trifluoromethyl)-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

56158-500MG 56158-1G 56158-5G 56158-25G
77329-584EA 225.57 USD
77329-584 77353-532 77365-874 77387-736
2-Chloro-4'-methyl-6-(trifluoromethyl)-1,1'-biphenyl ≥97%
2-Chloro-4'-methyl-6-(trifluoromethyl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₀ClF₃
MW: 270.677 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30469819
CAS Number: 2734778-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR