2,2-Dichloro-1-(3-fluoro-5-methoxyphenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

56135-1G 56135-5G 56135-25G
77341-624EA 96.82 USD
77341-624 77365-872 77377-776
2,2-Dichloro-1-(3-fluoro-5-methoxyphenyl)ethanol ≥95%
2,2-Dichloro-1-(3-fluoro-5-methoxyphenyl)ethanol

Precaución: for Research Use Only

Formula: C₉H₉Cl₂FO₂
MW: 239.071 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564343
CAS Number: 2734773-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR