(3-Bromo-5-chloro-4-ethoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

56114-500MG 56114-1G 56114-5G 56114-25G
77329-582EA 126.69 USD
77329-582 77344-138 77365-870 77377-768
(3-Bromo-5-chloro-4-ethoxyphenyl)methanol ≥97%
(3-Bromo-5-chloro-4-ethoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₀BrClO₂
MW: 265.531 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530485
CAS Number: 2385171-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR