5-(3-Chloro-4-methoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

56108-500MG 56108-1G 56108-5G
77329-580EA 338.87 USD
77329-580 77353-522 77377-766
5-(3-Chloro-4-methoxyphenyl)thiazol-2-amine ≥97%
5-(3-Chloro-4-methoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉ClN₂OS
MW: 240.709 g/mol
Storage Temperature: Ambient
CAS Number: 100988-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR