5-(4-Phenoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

56086-500MG 56086-1G 56086-5G
77329-578EA 340.93 USD
77329-578 77353-520 77377-764
5-(4-Phenoxyphenyl)thiazol-2-amine ≥97%
5-(4-Phenoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₅H₁₂N₂OS
Storage Temperature: Ambient
CAS Number: 1183879-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR